RIE156319271 Rieke Metals, Inc.

3,5-Dimethyl-1-adamantylzinc bromide

  • MDL number MFCD01311322
  • Cas Number : 312692-99-2
  • Molecular Weight : 308.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2062

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2062
Availability details for
Quantity
  • Availability
  • Add to Cart