RIE156319291 Rieke Metals, Inc.

4-Fluoro-1-naphthylmagnesium bromide

  • MDL number MFCD01311485
  • Cas Number : 17318-06-8
  • Molecular Weight : 249.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3034

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3034
Availability details for
Quantity
  • Availability
  • Add to Cart