RIE156319322 Rieke Metals, Inc.

2,5-Dimethylbenzylmagnesium chloride

  • MDL number MFCD01319906
  • Cas Number : 349552-22-3
  • Molecular Weight : 178.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  
Availability details for
Quantity
  • Availability
  • Add to Cart