RIE156319325 Rieke Metals, Inc.

2-Pyridylmagnesium bromide

  • MDL number MFCD01319909
  • Cas Number : 21970-13-8
  • Molecular Weight : 182.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3074

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3074
Availability details for
Quantity
  • Availability
  • Add to Cart