RIE156319329 Rieke Metals, Inc.

2,4-Dichlorobenzylmagnesium chloride

  • MDL number MFCD01319913
  • Cas Number : 129752-86-9
  • Molecular Weight : 219.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  
Availability details for
Quantity
  • Availability
  • Add to Cart