RIE156319337 Rieke Metals, Inc.

5-Acenaphthylmagnesium bromide

  • MDL number MFCD02260190
  • Cas Number : 620607-24-1
  • Molecular Weight : 257.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3093

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3093
Availability details for
Quantity
  • Availability
  • Add to Cart