RIE156319338 Rieke Metals, Inc.

3-Fluoro-4-biphenylmagnesium bromide

  • MDL number MFCD07698745
  • Cas Number : 76699-46-2
  • Molecular Weight : 275.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3094

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3094
Availability details for
Quantity
  • Availability
  • Add to Cart