RIE156319341 Rieke Metals, Inc.

2,4-Dimethylbenzylmagnesium chloride

  • MDL number MFCD02260194
  • Cas Number : 107549-24-6
  • Molecular Weight : 178.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3097

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3097
Availability details for
Quantity
  • Availability
  • Add to Cart