RIE156319346 Rieke Metals, Inc.

2,5-Dimethoxybenzylmagnesium chloride

  • MDL number MFCD02260199
  • Cas Number : 88185-17-5
  • Molecular Weight : 210.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3102

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3102
Availability details for
Quantity
  • Availability
  • Add to Cart