RIE156319357 Rieke Metals, Inc.

5-Cyano-5-methylhexylzinc bromide

  • MDL number MFCD01311419
  • Cas Number : 312693-22-4
  • Molecular Weight : 269.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2080

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2080
Availability details for
Quantity
  • Availability
  • Add to Cart