RIE156319365 Rieke Metals, Inc.

2-Methoxy-5-methylphenylmagnesium bromide

  • MDL number MFCD02260231
  • Cas Number : 70548-79-7
  • Molecular Weight : 225.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3135

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3135
Availability details for
Quantity
  • Availability
  • Add to Cart