RIE156319372 Rieke Metals, Inc.

4-iso-Propylphenylzinc iodide

  • MDL number MFCD01311424
  • Cas Number : 308796-20-5
  • Molecular Weight : 311.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/2085

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2085
Availability details for
Quantity
  • Availability
  • Add to Cart