RIE156319403 Rieke Metals, Inc.

4-(Thiomethyl)phenylzinc iodide

  • MDL number MFCD01863641
  • Cas Number : 307496-24-8
  • Molecular Weight : 315.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2107

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2107
Availability details for
Quantity
  • Availability
  • Add to Cart