RIE156319408 Rieke Metals, Inc.

3,4-(Ethylenedioxy)phenylmagnesium bromide

  • MDL number MFCD06200649
  • Cas Number : 107549-16-6
  • Molecular Weight : 239.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3188

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3188
Availability details for
Quantity
  • Availability
  • Add to Cart