RIE156319409 Rieke Metals, Inc.

4-tert-Butyl-2,6-dimethylphenylmagnesium bromide

  • MDL number MFCD06200650
  • Cas Number : 400656-18-0
  • Molecular Weight : 265.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3189

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3189
Availability details for
Quantity
  • Availability
  • Add to Cart