RIE156319417 Rieke Metals, Inc.

4-Ethylphenylzinc iodide

  • MDL number MFCD01311457
  • Cas Number : 312693-04-2
  • Molecular Weight : 297.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2118

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2118
Availability details for
Quantity
  • Availability
  • Add to Cart