RIE156319435 Rieke Metals, Inc.

2-Cyclohexylethylmagnesium bromide

  • MDL number MFCD08064089
  • Cas Number : 55766-17-1
  • Molecular Weight : 215.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/3216

Safety Information

MSDS http://riekemetals.com/products-services/catnum/3216
Availability details for
Quantity
  • Availability
  • Add to Cart