RIE156319459 Rieke Metals, Inc.

3,3'-Bithiophene

  • MDL number MFCD00041214
  • Cas Number : 3172-56-3
  • Molecular Weight : 166.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/4107

Safety Information

MSDS http://riekemetals.com/products-services/catnum/4107
Availability details for
Quantity
  • Availability
  • Add to Cart