RIE156319463 Rieke Metals, Inc.

5-oxo-5-(2-thienyl)valeric acid

  • MDL number MFCD00151848
  • Cas Number : 22971-62-6
  • Molecular Weight : 198.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/4115c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/4115c
Availability details for
Quantity
  • Availability
  • Add to Cart