RIE156319486 Rieke Metals, Inc.

2-Fluoro-4-methylphenylzinc bromide

  • MDL number MFCD02260132
  • Cas Number : 737797-14-7
  • Molecular Weight : 254.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2147

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2147
Availability details for
Quantity
  • Availability
  • Add to Cart