RIE156319490 Rieke Metals, Inc.

3-bromo-4'-cyanobenzophenone

  • MDL number MFCD00672020
  • Cas Number : 243137-97-5
  • Molecular Weight : 286.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5008

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5008
Availability details for
Quantity
  • Availability
  • Add to Cart