RIE156319496 Rieke Metals, Inc.

4-Carboethoxybenzophenone

  • MDL number MFCD00672025
  • Cas Number : 15165-27-2
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5014

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5014
Availability details for
Quantity
  • Availability
  • Add to Cart