RIE156319499 Rieke Metals, Inc.

4-Bromo-2'-carboethoxybenzophenone

  • MDL number MFCD00672028
  • Cas Number : 51476-11-0
  • Molecular Weight : 333.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5017

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5017
Availability details for
Quantity
  • Availability
  • Add to Cart