RIE156319500 Rieke Metals, Inc.

2-carboethoxybenzophenone

  • MDL number MFCD00027809
  • Cas Number : 604-61-5
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5018

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5018
Availability details for
Quantity
  • Availability
  • Add to Cart