RIE156319507 Rieke Metals, Inc.

4-bromo-4'-fluorobenzophenone

  • MDL number MFCD00672035
  • Cas Number : 2069-41-2
  • Molecular Weight : 279.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5025

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5025
Availability details for
Quantity
  • Availability
  • Add to Cart