RIE156319509 Rieke Metals, Inc.

cis-3-Carbomethoxycyclohexane-1-carboxylic acid

  • MDL number MFCD01311244
  • Cas Number : 733742-58-0
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/18505

Safety Information

MSDS http://riekemetals.com/products-services/catnum/18505
Availability details for
Quantity
  • Availability
  • Add to Cart