RIE156319512 Rieke Metals, Inc.

2,4-Difluorophenylzinc bromide

  • MDL number MFCD02260152
  • Cas Number : 517920-79-5
  • Molecular Weight : 258.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  
Availability details for
Quantity
  • Availability
  • Add to Cart