RIE156319513 Rieke Metals, Inc.

3,4'-difluorobenzophenone

  • MDL number MFCD00070811
  • Cas Number : 345-71-1
  • Molecular Weight : 218.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5035

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5035
Availability details for
Quantity
  • Availability
  • Add to Cart