RIE156319518 Rieke Metals, Inc.

3,3',4,5-tetrafluorobenzophenone

  • MDL number MFCD01319928
  • Cas Number : 746651-92-3
  • Molecular Weight : 254.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5041

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5041
Availability details for
Quantity
  • Availability
  • Add to Cart