RIE156319523 Rieke Metals, Inc.

6-Chlorohexylzinc bromide

  • MDL number MFCD00671973
  • Cas Number : 135579-83-8
  • Molecular Weight : 264.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2003

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2003
Availability details for
Quantity
  • Availability
  • Add to Cart