RIE156319526 Rieke Metals, Inc.

2-methoxybenzophenone

  • MDL number MFCD00017163
  • Cas Number : 2553-04-0
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5049-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5049-01
Availability details for
Quantity
  • Availability
  • Add to Cart