RIE156319531 Rieke Metals, Inc.

2-Chloro-4-fluorophenylzinc iodide

  • MDL number MFCD02260160
  • Cas Number : 497923-83-8
  • Molecular Weight : 321.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2173

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2173
Availability details for
Quantity
  • Availability
  • Add to Cart