RIE156319536 Rieke Metals, Inc.

3-Cyano-4-fluorophenylzinc bromide

  • MDL number MFCD02260165
  • Cas Number : 518990-00-6
  • Molecular Weight : 265.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2178

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2178
Availability details for
Quantity
  • Availability
  • Add to Cart