RIE156319543 Rieke Metals, Inc.

2,4-Dichlorophenylzinc iodide

  • MDL number MFCD02260174
  • Cas Number : 352530-42-8
  • Molecular Weight : 338.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2187

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2187
Availability details for
Quantity
  • Availability
  • Add to Cart