RIE156319555 Rieke Metals, Inc.

3-Thienylzinc iodide

  • MDL number MFCD03427251
  • Cas Number : 172035-85-7
  • Molecular Weight : 275.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2203

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2203
Availability details for
Quantity
  • Availability
  • Add to Cart