RIE156319574 Rieke Metals, Inc.

4-Carboethoxy-3'-methoxybenzophenone

  • MDL number MFCD01311549
  • Cas Number : 705279-41-0
  • Molecular Weight : 284.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5050-13

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-13
Availability details for
Quantity
  • Availability
  • Add to Cart