RIE156319578 Rieke Metals, Inc.

4-ethyl-3'-methoxybenzophenone

  • MDL number MFCD01311551
  • Cas Number : 82520-39-6
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-ethylphenyl)(3-methoxyphenyl)methanone
Inchi Code 1S/C16H16O2/c1-3-12-7-9-13(10-8-12)16(17)14-5-4-6-15(11-14)18-2/h4-11H,3H2,1-2H3
Inchi Key UDMMUFKUUJWDFE-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5050-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-17
Availability details for
Quantity
  • Availability
  • Add to Cart