RIE156319581 Rieke Metals, Inc.

3-chloro-3'-methoxybenzophenone

  • MDL number MFCD01311566
  • Cas Number : 32363-46-5
  • Molecular Weight : 246.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chlorophenyl)(3-methoxyphenyl)methanone
Inchi Code 1S/C14H11ClO2/c1-17-13-7-3-5-11(9-13)14(16)10-4-2-6-12(15)8-10/h2-9H,1H3
Inchi Key LBADCHNQRBNLEH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5050-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-20
Availability details for
Quantity
  • Availability
  • Add to Cart