RIE156319588 Rieke Metals, Inc.

2,5-dimethyl-3'-methoxybenzophenone

  • MDL number MFCD01311543
  • Cas Number : 750633-71-7
  • Molecular Weight : 240.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5050-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-28
Availability details for
Quantity
  • Availability
  • Add to Cart