RIE156319590 Rieke Metals, Inc.

3,4-dimethyl-3'-methoxybenzophenone

  • MDL number MFCD01311545
  • Cas Number : 750633-73-9
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dimethylphenyl)(3-methoxyphenyl)methanone
Inchi Code 1S/C16H16O2/c1-11-7-8-14(9-12(11)2)16(17)13-5-4-6-15(10-13)18-3/h4-10H,1-3H3
Inchi Key FWSSOWSGGCBGAU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5050-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5050-30
Availability details for
Quantity
  • Availability
  • Add to Cart