RIE156319591 Rieke Metals, Inc.

n-Octylzinc bromide

  • MDL number MFCD07698727
  • Cas Number : 131379-13-0
  • Molecular Weight : 258.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2224

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2224
Availability details for
Quantity
  • Availability
  • Add to Cart