RIE156319600 Rieke Metals, Inc.

4-methoxy-3'-methylbenzophenone

  • MDL number MFCD01311584
  • Cas Number : 53039-63-7
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5051-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5051-03
Availability details for
Quantity
  • Availability
  • Add to Cart