RIE156319627 Rieke Metals, Inc.

4-n-Pentylphenylzinc iodide

  • MDL number MFCD01311391
  • Cas Number : 308796-05-6
  • Molecular Weight : 339.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2040

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2040
Availability details for
Quantity
  • Availability
  • Add to Cart