RIE156319633 Rieke Metals, Inc.

2-fluoro-4'-methoxybenzophenone

  • MDL number MFCD00749331
  • Cas Number : 66938-29-2
  • Molecular Weight : 230.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5051-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5051-37
Availability details for
Quantity
  • Availability
  • Add to Cart