RIE156319634 Rieke Metals, Inc.

4,4'-Di-t-butylbenzophenone

  • MDL number MFCD01319937
  • Cas Number : 15796-82-4
  • Molecular Weight : 294.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5052

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5052
Availability details for
Quantity
  • Availability
  • Add to Cart