RIE156319635 Rieke Metals, Inc.

2,2'-Dinaphthyl ketone

  • MDL number MFCD00183496
  • Cas Number : 613-56-9
  • Molecular Weight : 282.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5053

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5053
Availability details for
Quantity
  • Availability
  • Add to Cart