RIE156319636 Rieke Metals, Inc.

4-t-butyl-4'-methylbenzophenone

  • MDL number MFCD01319938
  • Cas Number : 55709-38-1
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5054

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5054
Availability details for
Quantity
  • Availability
  • Add to Cart