RIE156319638 Rieke Metals, Inc.

2-naphthyl phenyl ketone

  • MDL number MFCD00004106
  • Cas Number : 644-13-3
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5056

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5056
Availability details for
Quantity
  • Availability
  • Add to Cart