RIE156319639 Rieke Metals, Inc.

3-Chlorophenylzinc iodide

  • MDL number MFCD01311393
  • Cas Number : 186000-42-0
  • Molecular Weight : 303.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2043

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2043
Availability details for
Quantity
  • Availability
  • Add to Cart