RIE156319647 Rieke Metals, Inc.

4-Bromobenzophenone ethylene ketal

  • MDL number MFCD02260358
  • Cas Number : 59793-76-9
  • Molecular Weight : 305.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5066

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5066
Availability details for
Quantity
  • Availability
  • Add to Cart